Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021788.2 | NP_068560.1 | 1 | 593 | ||
GenBank mRNA | AF075136.1 | - | 1 | 604 | ||
AK010928.1 | - | 1 | 594 | |||
AK088745.1 | - | 1 | 598 | |||
BC132081.1 | - | 1 | 576 | 40130358 | ||
BC132087.1 | - | 1 | 576 | 40130364 |