Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jarid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205043.1 | NP_001191972.1 | 2, 3 | 780 | ||
NM_001205044.1 | NP_001191973.1 | 3, 4 | 453 | |||
NM_021878.3 | NP_068678.1 | 3, 4 | 499 | |||
GenBank mRNA | AK044888.1 | - | 2, 3 | 684 | ||
AK045214.1 | - | 2, 3 | 585 | |||
AK147226.1 | - | 2, 3 | 780 | |||
BC026056.1 | - | 2, 3 | 687 | |||
BC050129.1 | - | 2, 3 | 701 | |||
BC051010.1 | - | 2, 3 | 683 | |||
BC052444.1 | - | 3, 4 | 370 | 6406875 | ||
BC060695.1 | - | 3, 4 | 370 | 6406875 | ||
D31967.1 | - | 3, 4 | 389 |