Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021882.4 | NP_068682.2 | 3 | 239 | ||
GenBank mRNA | AK012808.1 | - | 3 | 240 | ||
AK131956.1 | - | 3 | 241 | |||
BC075667.1 | - | 3 | 226 | |||
BC082555.1 | - | 3 | 225 | 30541702 | ||
U14133.1 | - | 3 | 214 |