Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021887.2 | NP_068687.1 | 7, 8 | 1228 | ||
GenBank mRNA | AB049137.1 | - | 7, 8 | 1120 | ||
AF254068.1 | - | 7, 8 | 932 | |||
AF269134.1 | - | 6, 7 | 790 | |||
AF279436.1 | - | 7, 8 | 1180 | |||
AF477982.1 | - | 6, 7 | 806 | |||
AF477983.1 | - | 6, 7 | 806 | |||
AF477984.1 | - | 6, 7 | 806 | |||
AF477985.1 | - | 6, 7 | 806 | |||
AF477986.1 | - | 6, 7 | 806 | |||
AK150824.1 | - | 7, 8 | 1189 | |||
AK171826.1 | - | 7, 8 | 1167 | |||
AK172032.1 | - | 7, 8 | 1200 |