Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021889.4 | NP_068689.2 | 3 | 971 | ||
GenBank mRNA | AB026802.1 | - | 3 | 770 | ||
AK089115.1 | - | 3 | 972 | |||
AK156631.1 | - | 3 | 972 | |||
BC132495.1 | - | 3 | 958 | 40130772 | ||
BC137904.1 | - | 3 | 958 | 8860923 | ||
BC145024.1 | - | 3 | 958 |