Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021896.5 | NP_068696.2 | 3, 4 | 635 | ||
GenBank mRNA | AF297082.1 | - | 3, 4 | 610 | ||
AK004815.1 | - | 3, 4 | 569 | |||
AK028558.1 | - | 3, 4 | 558 | |||
AK031305.1 | - | 3, 4 | 548 | |||
BC057327.1 | - | 3, 4 | 545 | 6825960 | ||
BC094310.1 | - | 3, 4 | 691 | 6838836 |