Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022004.6 | NP_071287.1 | 5 | 493 | ||
GenBank mRNA | AB032010.2 | - | 5 | 206 | ||
AK003888.1 | - | 5 | 256 | |||
AK018354.1 | - | 5 | 226 | |||
BC051127.1 | - | 5 | 264 | 6531451 | ||
BQ891313.1 | - | 5 | 207 | |||
BY704145.1 | - | 5 | 250 | |||
W34202.1 | - | 5 | 213 |