Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146001.1 | NP_001139473.1 | 5, 6 | 849 | ||
NM_021517.2 | NP_067492.2 | 5, 6 | 895 | |||
GenBank mRNA | AF220100.1 | - | 5, 6 | 834 | ||
AF474247.1 | - | 4, 5 | 833 | |||
AK006269.1 | - | 5, 6 | 1369 | |||
AK012070.1 | - | 5, 6 | 885 | |||
AK014879.1 | - | 5, 6 | 1152 | |||
AK029752.1 | - | 5, 6 | 1353 | |||
AK029764.1 | - | 5, 6 | 1363 | |||
AK165415.1 | - | 5, 6 | 830 | |||
BC013512.1 | - | 4, 5 | 818 | 4234949 |