Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021540.4 | NP_067515.2 | 8, 9 | 1323 | ||
GenBank mRNA | AF171875.1 | - | 8, 9 | 1291 | ||
AK191139.1 | - | 2, 3 | 211 | |||
BC018199.1 | - | 8, 9 | 1317 | 4017435 | ||
BC048901.1 | - | 8, 9 | 1306 | 6390079 |