Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021542.4 | NP_067517.1 | 5 | 1415 | ||
GenBank mRNA | AF259395.1 | - | 5 | 1085 | ||
AF319542.1 | - | 5 | 1154 | |||
AK085330.1 | - | 5 | 1416 | |||
AK132172.1 | - | 5 | 1443 | |||
AK172608.1 | - | 5 | 1416 | |||
BC034012.1 | - | 5 | 1417 | 4239113 | ||
BC058164.1 | - | 5 | 1406 | 5323480 |