Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042726.3 | NP_001036191.1 | 3 | 3100 | ||
NM_021543.4 | NP_067518.2 | 3 | 3391 | |||
GenBank mRNA | AF231125.1 | - | 3 | 3315 | ||
AK048638.1 | - | 3 | 3101 | |||
AK049982.1 | - | 1 | 3934 | |||
AK140407.1 | - | 3 | 2988 | |||
BC052388.1 | - | 1 | 3910 | 5718846 | ||
BC053008.1 | - | 3 | 3379 | 5698009 |