Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253860.1 | NP_001240789.1 | 22 | 4262 | ||
NM_021544.4 | NP_067519.2 | 23 | 4430 | |||
GenBank mRNA | AJ271477.1 | - | 22 | 4255 | ||
AK147254.1 | - | 23 | 4430 | |||
AK147517.1 | - | 23 | 4430 |