Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021554.2 | NP_067529.2 | 5 | 1269 | ||
GenBank mRNA | AB041664.1 | - | 5 | 1046 | ||
AJ278576.1 | - | 5 | 1269 | |||
AK003378.1 | - | 1 | 80 | |||
AK075586.1 | - | 5 | 1031 | |||
AK084366.1 | - | 995 | ||||
AK169951.1 | - | 4 | 685 | |||
AK206642.1 | - | 1 | 55 | |||
AK210143.1 | - | 1 | 55 | |||
AK211126.1 | - | 1 | 55 | |||
AK211493.1 | - | 1 | 55 | |||
AK215069.1 | - | 1 | 55 | |||
AK216352.1 | - | 1 | 55 | |||
AK216736.1 | - | 2 | 347 | |||
BC023188.1 | - | 4 | 710 | |||
BC068124.1 | - | 5 | 1063 | 5716972 | ||
BC082317.1 | - | 4 | 694 |