Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021567.5 | NP_067542.3 | 6 | 599 | ||
GenBank mRNA | AF176328.1 | - | 6 | 485 | ||
AK004580.1 | - | 6 | 550 | |||
AK005383.1 | - | 6 | 548 | |||
AK077632.1 | - | 6 | 549 | |||
AK082005.1 | - | 6 | 549 | |||
AK178757.1 | - | 2 | 218 | |||
AK179721.1 | - | 2 | 219 | |||
AK179793.1 | - | 2 | 137 | |||
AK179802.1 | - | 2 | 218 | |||
AK187694.1 | - | 2 | 218 | |||
AK188206.1 | - | 2 | 219 | |||
AK188263.1 | - | 2 | 218 | |||
AK189619.1 | - | 2 | 218 | |||
BC010694.1 | - | 6 | 488 | 2654291 |