Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018053.1 | NP_001018063.1 | 6 | 524 | ||
NM_006212.2 | NP_006203.2 | 6 | 524 | |||
GenBank mRNA | AB044805.1 | - | 6 | 477 | ||
AF470623.1 | - | 6 | 496 | |||
AJ005578.1 | - | 6 | 501 | |||
AK292883.1 | - | 6 | 549 | |||
AK302315.1 | - | 8 | 760 | |||
BC069350.1 | - | 6 | 465 | 7262370 | ||
BC069385.1 | - | 6 | 465 | 7262287 | ||
BC069583.1 | - | 6 | 465 | 7262299 | ||
BC069586.1 | - | 6 | 465 | 7262311 | ||
BC069600.1 | - | 6 | 465 | 7262323 | ||
BC075075.2 | - | 6 | 462 | 30915381 | ||
BC075076.2 | - | 6 | 462 | 30915594 | ||
BC112103.1 | - | 6 | 467 | 8327571 | ||
BC112105.1 | - | 6 | 467 | 8327573 | ||
EU334168.1 | - | 6 | 542 |