Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021605.3 | NP_067618.1 | 10 | 2183 | ||
GenBank mRNA | AF217650.1 | - | 9 | 2036 | ||
AK035502.1 | - | 10 | 2182 | |||
AK088173.1 | - | 10 | 2232 | |||
AK165583.1 | - | 8 | 1952 | |||
AK169649.1 | - | 1 | 1019 |