Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC125528.1 | - | 4, 5 | 493 | ||
BY714594.1 | - | 1, 2 | 180 | |||
CJ236701.1 | - | 4, 5 | 310 | |||
X65979.1 | - | 3, 4 | 318 | |||
X65981.1 | - | 5, 6 | 615 |