Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021791.3 | NP_068563.2 | 9, 10 | 1021 | ||
GenBank mRNA | AB046665.1 | - | 8, 9 | 1019 | ||
BC061104.1 | - | 9, 10 | 1021 | 30296398 |