Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021886.1 | NP_068686.1 | 9 | 1155 | ||
GenBank mRNA | AB017634.1 | - | 9 | 1155 | ||
AK006208.1 | - | 9 | 1139 | |||
AK010533.1 | - | 6 | 1217 | |||
AK011264.1 | - | 9 | 1141 | |||
AK011738.1 | - | 9 | 1139 | |||
AK012907.1 | - | 9 | 1100 |