Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021887.2 | NP_068687.1 | 9 | 2506 | ||
GenBank mRNA | AF279436.1 | - | 9 | 2482 | ||
AK137793.1 | - | 6 | 3422 |