Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199105.1 | NP_001186034.1 | 5 | 2758 | ||
NM_021897.3 | NP_068697.1 | 4 | 2720 | |||
GenBank mRNA | AJ131776.1 | - | 4 | 2721 | ||
AY034611.1 | - | 5 | 2759 | |||
AY034612.1 | - | 4 | 2721 |