Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022015.3 | NP_071298.1 | 6 | 635 | ||
GenBank mRNA | AF222802.1 | - | 6 | 637 | ||
AK045108.1 | - | 6 | 636 | |||
AK049949.1 | - | 6 | 637 | |||
AK076486.1 | - | 6 | 635 | |||
AK086656.1 | - | 6 | 636 | |||
AK161658.1 | - | 6 | 636 | |||
AK162110.1 | - | 6 | 636 | |||
BC023756.1 | - | 6 | 640 | |||
BC057895.1 | - | 6 | 622 | 5031867 |