Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022017.3 | NP_071300.2 | 2 | 248 | ||
GenBank mRNA | AB021875.2 | - | 1 | 175 | ||
AF208026.1 | - | 2 | 216 | |||
AF279672.1 | - | 2 | 232 | |||
AJ296078.1 | - | 1 | 167 | |||
AK002735.1 | - | 2 | 249 | |||
AK083223.1 | - | 2 | 218 | |||
BC127052.1 | - | 2 | 213 | 40109678 |
Gene Symbol: Coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025633.1 | - | 205 |