Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022019.5 | NP_071302.2 | 2 | 419 | ||
GenBank mRNA | AB037908.1 | - | 2 | 425 | ||
AK011995.1 | - | 2 | 403 | |||
AK035293.1 | - | 2 | 417 | |||
AK050528.1 | - | 2 | 417 | |||
AK088024.1 | - | 2 | 418 | |||
AK088186.1 | - | 2 | 420 | |||
AK088357.1 | - | 2 | 415 | |||
AK142568.1 | - | 2 | 421 | |||
BC025066.1 | - | 2 | 437 | 3495358 |