Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022025.4 | NP_071308.2 | 3 | 710 | ||
GenBank mRNA | AF276872.2 | - | 2 | 201 | ||
AJ401467.1 | - | 2 | 201 | |||
AK034415.1 | - | 3 | 594 | |||
AK053063.1 | - | 3 | 712 | |||
BC065089.1 | - | 2 | 227 | 6851099 |