Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163434.1 | NP_001156906.1 | 2 | 224 | ||
NM_022310.3 | NP_071705.3 | 1 | 306 | |||
GenBank mRNA | AJ002387.1 | - | 1 | 85 | ||
AK004578.1 | - | 1 | 301 | |||
AK076079.1 | - | 2 | 220 | |||
AK077463.1 | - | 2 | 224 | |||
AK144650.1 | - | 2 | 220 | |||
AK144742.1 | - | 2 | 220 | |||
AK146647.1 | - | 2 | 223 | |||
AK148539.1 | - | 2 | 223 | |||
AK149447.1 | - | 2 | 223 | |||
AK151244.1 | - | 1 | 172 | |||
AK151647.1 | - | 2 | 222 | |||
AK151806.1 | - | 2 | 223 | |||
AK152020.1 | - | 2 | 220 | |||
AK152521.1 | - | 1 | 303 | |||
AK152958.1 | - | 2 | 223 | |||
AK159719.1 | - | 2 | 220 | |||
AK160821.1 | - | 224 | ||||
AK166207.1 | - | 2 | 220 | |||
AK166739.1 | - | 2 | 220 | |||
AK167011.1 | - | 2 | 220 | |||
AK168012.1 | - | 2 | 224 | |||
AK168230.1 | - | 2 | 219 | |||
AK169034.1 | - | 2 | 224 | |||
AK197258.1 | - | 1 | 22 | |||
AK215673.1 | - | 1 | 22 | |||
BC050927.1 | - | 1 | 283 | 6415084 | ||
D78645.1 | - | 1 | 166 |