Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022319.2 | NP_071714.2 | 3 | 485 | ||
GenBank mRNA | AJ278069.1 | - | 3 | 433 | ||
AK220500.1 | - | 3 | 499 | |||
BC063058.1 | - | 3 | 485 | 5705591 |