Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162500.1 | NP_001155972.1 | 2, 3 | 261 | ||
NM_022321.4 | NP_071716.3 | 2, 3 | 216 | |||
GenBank mRNA | AF312712.1 | - | 2, 3 | 174 | ||
AK028329.1 | - | 2, 3 | 217 | |||
AK037365.1 | - | 2, 3 | 188 | |||
AK040914.1 | - | 2, 3 | 147 | |||
AK079881.1 | - | 2, 3 | 142 | |||
AK088419.1 | - | 2, 3 | 166 | |||
BC011200.1 | - | 2, 3 | 170 | 4190510 | ||
BY200543.1 | - | 2, 3 | 262 |