Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282684.1 | NP_001269613.1 | 6 | 726 | ||
NM_024681.3 | NP_078957.2 | 6 | 726 | |||
XM_005261741.3 | XP_005261798.2 | 6 | 737 | |||
XM_005261742.2 | XP_005261799.2 | 6 | 737 | |||
XM_005261743.2 | XP_005261800.2 | 6 | 737 | |||
XM_005261744.2 | XP_005261801.1 | 6 | 737 | |||
GenBank mRNA | AK022304.1 | - | 6 | 702 | ||
AK308720.1 | - | 6 | 975 | |||
BC025403.1 | - | 6 | 725 | 4824153 | ||
BC031038.1 | - | 6 | 709 | 5277700 |