Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022328.2 | NP_071723.1 | 2 | 272 | ||
GenBank mRNA | AF298887.1 | - | 1 | 158 | ||
AF312858.1 | - | 2 | 169 | |||
AK144605.1 | - | 2 | 300 | |||
AK156820.1 | - | 2 | 299 | |||
BC053005.1 | - | 2 | 272 | 5716396 |