Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022424.5 | NP_071869.2 | 7 | 1115 | ||
GenBank mRNA | AB030187.1 | - | 3 | 287 | ||
AK018154.1 | - | 7 | 1111 | |||
AK163183.1 | - | 7 | 1116 | |||
BC027164.1 | - | 3 | 269 | |||
BC139278.1 | - | 7 | 867 | 8862300 | ||
BC139292.1 | - | 7 | 867 | 8862314 |