Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022656.2 | NP_073147.2 | 10 | 1223 | ||
GenBank mRNA | AF315344.1 | - | 6 | 658 | ||
AK004628.1 | - | 10 | 1060 | |||
AK036043.1 | - | 10 | 1078 | |||
AK049791.1 | - | 10 | 1078 | |||
AK160990.1 | - | 10 | 1097 | |||
BC003270.1 | - | 10 | 1085 | |||
BC066838.1 | - | 10 | 1081 | |||
BC089329.1 | - | 5 | 442 | |||
BC108364.1 | - | 5 | 418 | |||
BC144980.1 | - | 6 | 552 |