Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022657.4 | NP_073148.1 | 1 | 279 | ||
GenBank mRNA | AB037889.1 | - | 1 | 151 | ||
AF263536.1 | - | 1 | 207 | |||
AK155687.1 | - | 1 | 279 | |||
BC120605.1 | - | 1 | 174 | 40129528 |