Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285800.1 | NP_001272729.1 | 1 | 145 | ||
NM_022811.3 | NP_073722.1 | 1 | 145 | |||
GenBank mRNA | AK050780.1 | - | 93 | |||
AK083948.1 | - | 1 | 116 | |||
AK132556.1 | - | 1 | 116 | |||
BC034783.1 | - | 1 | 92 | 4022372 | ||
BY328290.1 | - | 1 | 147 | |||
D14336.1 | - | 1 | 108 |