Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022814.2 | NP_073725.2 | 2 | 948 | ||
GenBank mRNA | AF206329.1 | - | 2 | 948 | ||
AK035333.1 | - | 2 | 966 | |||
AK045217.1 | - | 2 | 1041 | |||
AK052699.1 | - | 2 | 920 | |||
AK087058.1 | - | 2 | 993 |