Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 6 | 641 | ||
NM_001199114.1 | NP_001186043.1 | 5 | 480 | |||
NM_001199115.1 | NP_001186044.1 | 5 | 586 | |||
NM_001199116.1 | NP_001186045.1 | 5 | 546 | |||
NM_022880.3 | NP_075018.1 | 4 | 587 | |||
GenBank mRNA | AF131212.1 | - | 4 | 588 | ||
AF305501.1 | - | 5 | 551 | |||
AK004756.1 | - | 5 | 532 | |||
AK050089.1 | - | 5 | 571 | |||
AK146265.1 | - | 5 | 481 | |||
AK149483.1 | - | 5 | 502 | |||
AK170463.1 | - | 5 | 518 | |||
AK181560.1 | - | 2 | 126 | |||
AK193260.1 | - | 1 | 21 | |||
AK214211.1 | - | 1 | 21 | |||
BC004828.1 | - | 6 | 604 | 2631728 | ||
BC006812.1 | - | 4 | 518 | 3155188 | ||
CD540408.1 | - | 4 | 506 | |||
CX219686.1 | - | 6 | 636 | |||
EU180577.1 | - | 4 | 456 |