Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164885.1 | NP_001158357.1 | 2, 3 | 420 | ||
NM_022882.4 | NP_075020.2 | 2, 3 | 296 | |||
GenBank mRNA | AF286723.1 | - | 2, 3 | 296 | ||
AK033662.1 | - | 2, 3 | 333 | |||
AK050018.1 | - | 2, 3 | 421 | |||
AK086834.1 | - | 2, 3 | 232 | |||
AK136472.1 | - | 891 | ||||
BC039698.1 | - | 2, 3 | 409 | 5101211 |