Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022889.3 | NP_075027.1 | 3 | 194 | ||
GenBank mRNA | AF289539.1 | - | 3 | 154 | ||
AK029075.1 | - | 3 | 194 | |||
AK088831.1 | - | 3 | 172 | |||
AK220142.1 | - | 3 | 141 | |||
BC004844.1 | - | 3 | 179 | 2649598 | ||
BC011142.1 | - | 3 | 177 | 4207164 |