Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038699.2 | NP_001033788.1 | 2 | 269 | ||
NM_022014.4 | NP_071297.1 | 2 | 269 | |||
GenBank mRNA | AA058133.1 | - | 2 | 225 | ||
AJ404616.1 | - | 2 | 255 | |||
AK010151.1 | - | 2 | 263 | |||
AK030362.1 | - | 2 | 256 | |||
AK172235.1 | - | 2 | 254 | |||
BC054374.1 | - | 2 | 250 | 6313660 | ||
BC064710.1 | - | 2 | 255 | 30380196 | ||
BY142714.1 | - | 2 | 271 |