Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Perp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022032.4 | NP_071315.1 | 3 | 759 | ||
GenBank mRNA | AF249870.1 | - | 3 | 760 | ||
AK003734.1 | - | 3 | 700 | |||
AK009184.1 | - | 3 | 697 | |||
AK009190.1 | - | 3 | 698 | |||
BC021772.1 | - | 3 | 684 | 3985702 |