Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163434.1 | NP_001156906.1 | 3 | 491 | ||
NM_022310.3 | NP_071705.3 | 2 | 573 | |||
GenBank mRNA | AJ002387.1 | - | 2 | 352 | ||
AK004578.1 | - | 2 | 568 | |||
AK076079.1 | - | 3 | 487 | |||
AK077463.1 | - | 3 | 491 | |||
AK144650.1 | - | 3 | 487 | |||
AK144742.1 | - | 3 | 487 | |||
AK146647.1 | - | 3 | 490 | |||
AK148539.1 | - | 3 | 490 | |||
AK149447.1 | - | 3 | 490 | |||
AK151244.1 | - | 2 | 439 | |||
AK151647.1 | - | 3 | 489 | |||
AK151806.1 | - | 3 | 490 | |||
AK152020.1 | - | 3 | 487 | |||
AK152521.1 | - | 2 | 570 | |||
AK152958.1 | - | 3 | 490 | |||
AK159719.1 | - | 3 | 487 | |||
AK160821.1 | - | 491 | ||||
AK166207.1 | - | 3 | 487 | |||
AK166739.1 | - | 3 | 487 | |||
AK167011.1 | - | 3 | 487 | |||
AK168012.1 | - | 3 | 491 | |||
AK168230.1 | - | 3 | 486 | |||
AK169034.1 | - | 3 | 491 | |||
AK178434.1 | - | 1 | 85 | |||
AK178841.1 | - | 1 | 86 | |||
AK181293.1 | - | 1 | 85 | |||
AK188133.1 | - | 1 | 85 | |||
AK188254.1 | - | 1 | 85 | |||
AK192496.1 | - | 1 | 82 | |||
AK195133.1 | - | 1 | 47 | |||
AK197130.1 | - | 1 | 38 | |||
AK198934.1 | - | 1 | 46 | |||
AK200033.1 | - | 1 | 45 | |||
BC005785.1 | - | 1 | 207 | |||
BC050927.1 | - | 2 | 550 | 6415084 | ||
D78645.1 | - | 2 | 433 | |||
U89425.1 | - | 1 | 35 |