Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022430.2 | NP_071875.2 | 5 | 622 | ||
GenBank mRNA | AB026044.1 | - | 5 | 572 | ||
AF237906.1 | - | 5 | 560 | |||
AK008099.1 | - | 5 | 561 | |||
AY039038.1 | - | 5 | 560 | |||
BC042719.1 | - | 5 | 608 |