Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122765.1 | NP_001116237.1 | 3 | 257 | ||
NM_022432.4 | NP_071877.3 | 4 | 304 | |||
GenBank mRNA | AB221570.1 | - | 4 | 209 | ||
AF299337.1 | - | 4 | 289 | |||
AK014042.1 | - | 3 | 228 | |||
AK144431.1 | - | 3 | 200 | |||
AK146412.1 | - | 3 | 237 | |||
AK193014.1 | - | 3 | 216 | |||
AK203445.1 | - | 2 | 147 | |||
BB873718.1 | - | 3 | 257 | |||
BC021439.1 | - | 4 | 308 | 5039278 | ||
KF032392.1 | - | 4 | 246 |