Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022653.4 | NP_073144.3 | 3 | 424 | ||
GenBank mRNA | AF314187.1 | - | 3 | 382 | ||
AK028609.1 | - | 3 | 358 | |||
BC031722.1 | - | 3 | 390 | 4019169 | ||
CF163503.1 | - | 3 | 405 |