Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271472.1 | NP_001258401.1 | 2, 3 | 350 | ||
NM_001271473.1 | NP_001258402.1 | 2, 3 | 350 | |||
NM_022723.3 | NP_073560.2 | 2, 3 | 350 | |||
GenBank mRNA | AF276425.1 | - | 2, 3 | 217 | ||
AK041590.1 | - | 2, 3 | 333 | |||
BC066066.1 | - | 2, 3 | 308 | 6850999 | ||
BC080278.1 | - | 2, 3 | 318 | 30354974 | ||
CJ104466.1 | - | 2, 3 | 351 |