Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022885.2 | NP_075023.2 | 10 | 1366 | ||
GenBank mRNA | AF233321.1 | - | 3 | 385 | ||
AF461761.1 | - | 10 | 1265 | |||
AK051183.1 | - | 10 | 1366 | |||
AK088004.1 | - | 9 | 1110 | |||
AK090176.1 | - | 10 | 1275 | |||
AK166006.1 | - | 10 | 1176 | |||
AK180687.1 | - | 2 | 97 | |||
BC029033.1 | - | 7 | 808 | |||
BC033452.1 | - | 10 | 1265 | 3156622 |