Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022018.3 | NP_071301.2 | 14 | 2268 | ||
GenBank mRNA | AB049355.1 | - | 14 | 2108 | ||
AK029847.1 | - | 10 | 1580 | |||
AK136597.1 | - | 14 | 2178 | |||
AK170193.1 | - | 15 | 2346 | |||
AK170347.1 | - | 5 | 863 | |||
BC058234.1 | - | 12 | 1868 | 6306849 | ||
BC099690.1 | - | 14 | 2175 | |||
BC137843.1 | - | 10 | 1645 | 8860862 |