Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104529.1 | NP_001097999.1 | 2 | 757 | ||
NM_001271766.1 | NP_001258695.1 | 2 | 696 | |||
NM_022320.4 | NP_071715.3 | 2 | 993 | |||
GenBank mRNA | AF200349.1 | - | 2 | 678 | ||
AK034870.1 | - | 2 | 777 | |||
AK036503.1 | - | 2 | 755 | |||
AK089198.1 | - | 2 | 1011 | |||
AK154203.1 | - | 2 | 680 | |||
AK170995.1 | - | 2 | 697 | |||
BC027429.1 | - | 2 | 630 | 3669814 |