Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160180.1 | NP_001153652.1 | 3 | 1304 | ||
NM_001160181.1 | NP_001153653.1 | 3 | 1252 | |||
NM_001160182.1 | NP_001153654.1 | 3 | 1183 | |||
NM_022329.4 | NP_071724.1 | 2 | 1192 | |||
GenBank mRNA | AJ251363.1 | - | 2 | 1159 | ||
AK003852.1 | - | 2 | 1141 | |||
AK032697.1 | - | 2 | 1136 | |||
AK075980.1 | - | 1 | 423 | |||
AK150117.1 | - | 3 | 1307 | |||
AK150303.1 | - | 3 | 1221 | |||
AK152137.1 | - | 3 | 1228 | |||
BC039927.1 | - | 2 | 1157 | |||
BC053726.1 | - | 3 | 1207 | |||
BC060258.1 | - | 3 | 1533 | |||
BC064017.1 | - | 2 | 1157 | 3995710 | ||
BC092059.1 | - | 3 | 1210 | 4019181 |