Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022411.3 | NP_071856.1 | 3 | 278 | ||
GenBank mRNA | AF201903.1 | - | 3 | 278 | ||
AK140972.1 | - | 3 | 328 | |||
BC013493.1 | - | 3 | 303 | 4218337 |